missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Porcine IL 1RA Protein

A cDNA sequence encoding the IL 1RA was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10698-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15934688

  • £2975.00 / 1mg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

IL 1RA Protein
Research Use Only
The ED50 as determined by inhibiting IL-1a-dependent proliferation of murine D10S cells is less than 50 ng/ml, corresponding to a specific activity of > 2.0 M 104 IU/mg in the presence of 75 pg/ml rPoIL-1a.
Porcine
Untagged
The IL 1RA was lyophilized from a 0.2microm filtered concentrated solution in 1M PBS, pH 7.4, containing 1mM DTT.
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
HPLGKRPCRM QAFRIWDVNQ KTFYLRNNQL VAGYLQGPNT KLEEKIDVVP VEPHFVFLGI HGGKLCLSCV KSGDEMKLQL DAVNITDLRK NSEQDKRFTF IRSDSGPTTS FESAACPGWF LCTALEADQP VGLTNTPKAA VKVTKFYFQQ DQ
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers