missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Porcine IL 1 alpha Protein
A cDNA sequence encoding the IL 1 alpha was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10685-10ug
Additional Details : Weight : 0.01000kg
Specifications
397094 | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MAKVPDLFED LKNCYSENEE YSSDIDHLSL NQKSFYDASY EPLPGDGMDK FMPLSTSKTS KTSRLNFKDS VVMAAANGKI LKKRRLSLNQ FITDDDLEAI ANDTEEEIIK PRSATYSFQS NMKYNFMRVI NHQCILNDAR NQSIIRDPSG QYLMAAVLNN LDEAVKFDMA AYTSNDDSQL PVTLRISETR LFVSAQNEDE PVLLKELPET PKTIKDETSL LFFWEKHGNM DYFKSAAHPK LFIATRQEKL VHMAPGLPSV TDFQILENQS | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
IL 1 alpha Protein | |
Research Use Only | |
IL1A | |
Porcine | |
Untagged | |
The protein was lyophilized from a concentrated (1 mg/ml) sterile solution containing no additives. |