missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Porcine IL 1 alpha Protein

A cDNA sequence encoding the IL 1 alpha was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10685-0.1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15944648

  • £1435.00 / 100µg
Estimated Shipment: 09-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

397094
100 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MAKVPDLFED LKNCYSENEE YSSDIDHLSL NQKSFYDASY EPLPGDGMDK FMPLSTSKTS KTSRLNFKDS VVMAAANGKI LKKRRLSLNQ FITDDDLEAI ANDTEEEIIK PRSATYSFQS NMKYNFMRVI NHQCILNDAR NQSIIRDPSG QYLMAAVLNN LDEAVKFDMA AYTSNDDSQL PVTLRISETR LFVSAQNEDE PVLLKELPET PKTIKDETSL LFFWEKHGNM DYFKSAAHPK LFIATRQEKL VHMAPGLPSV TDFQILENQS
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
IL 1 alpha Protein
Research Use Only
IL1A
Porcine
Untagged
The protein was lyophilized from a concentrated (1 mg/ml) sterile solution containing no additives.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers