missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Porcine IL 1 alpha Protein
A cDNA sequence encoding the IL 1 alpha was constructed and used to recombinantly synthesize the protein.
£99.15 - £1435.00
Specifications
Gene ID (Entrez) | 397094 |
---|---|
Name | IL 1 alpha Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | IL1A |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15964648
|
enQuireBio™
QP10685-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15954648
|
enQuireBio™
QP10685-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15944648
|
enQuireBio™
QP10685-0.1MG |
100 μg |
£1435.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
397094 | |
Research Use Only | |
IL1A | |
Porcine | |
Untagged | |
The protein was lyophilized from a concentrated (1 mg/ml) sterile solution containing no additives. |
IL 1 alpha Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MAKVPDLFED LKNCYSENEE YSSDIDHLSL NQKSFYDASY EPLPGDGMDK FMPLSTSKTS KTSRLNFKDS VVMAAANGKI LKKRRLSLNQ FITDDDLEAI ANDTEEEIIK PRSATYSFQS NMKYNFMRVI NHQCILNDAR NQSIIRDPSG QYLMAAVLNN LDEAVKFDMA AYTSNDDSQL PVTLRISETR LFVSAQNEDE PVLLKELPET PKTIKDETSL LFFWEKHGNM DYFKSAAHPK LFIATRQEKL VHMAPGLPSV TDFQILENQS | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |