missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Porcine FSH Protein

A cDNA sequence encoding the FSH was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11904-5000IU

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15968748

  • £431.40 / 5000 units
Estimated Shipment: 12-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

FSH Protein
Research Use Only
Recombinant Protein
Native Protein
MKSLQFCFLF CCWKAICCNS CELTNITITV EKEECNFCIS INTTWCAGYC YTRDLVYKDP ARPNIQKTCT FKELVYETVK VPGCAHHADS LYTYPVATEC HCGKCDSDST DCTVRGLGPS YCSFSEMKE
5000 IU
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Porcine
Untagged
The FSH was lyophilized with no additives.
Videos
SDS
Documents
Certificates
Special Offers

Special Offers