missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Porcine CD163 Protein
A cDNA sequence encoding the CD163 was constructed and used to recombinantly synthesize the protein.
£99.15 - £1985.00
Specifications
Gene ID (Entrez) | 397031 |
---|---|
Name | CD163 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | CD163 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15926778
|
enQuireBio™
QP11303-10UG |
10 μg |
£99.15
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15946778
|
enQuireBio™
QP11303-50UG |
50 μg |
£155.00
50µg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15936778
|
enQuireBio™
QP11303-1MG |
1 mg |
£1985.00
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
397031 | |
Research Use Only | |
CD163 | |
Porcine | |
His | |
The protein was lyophilized from a 0.2microm filtered concentrated solution in 1M PBS, pH 7.4, containing 4M Urea. |
CD163 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MDKLRMVLHE NSGSADLKLR VVDGVTECSG RLEVKFQGEW GTICDDGWDS DDAAVACKQL GCPTAVTAIG RVNASEGTGH IWLDSVSCHG HESALWQCRH HEWGKHYCNH NEDAGVTCSD GSDLELRLKG GGSHCAGTVE VEIQKLVGKV CDRSWGLKEA DVVCRQLGCG SALKTSYQVY SKTKATNTWL FVSSCNGNET SLWDCKNWQW GGLSCDHYDE AKITCSAHRK PRLVGGDIPC SGRVEVQHGD TWGTVCDSDF SLEAASVLCR ELQCGTVVSL LGGAHFGEGS GQIWAEEFQC EGHESHLSLC PVAPRPDGTC SHSRDVGVVC SRYTQIRLVN GKTPCEGRVE LNILGSWGSL CNSHWDMEDA HVLCQQLKCG VALSIPGGAP FGKGSEQVWR HMFHCTGTEK HMGDCSVTAL GASLCSSGQV ASVICSGNQS QTLSPCNSSS SDPSSSIISE ENGVACIGSG QLRLVDGGGR CAGRVEVYHE GSWGTICDDS WDLNDAHVVC KQLSCGWAIN ATGSAHFGEG TGPIWLDEIN CNGKESHIWQ CHSHGWGRHN CRHKEDAGVI CSEFMSLRLI SENSRETCAG RLEVFYNGAW GSVGKNSMSP ATVGVVCRQL GCADRGDISP ASSDKTVSRH MWVDNVQCPK GPDTLWQCPS SPWKKRLASP SEETWITCAN KIRLQEGNTN CSGRVEIWYG GSWGTVCDDS WDLEDAQVVC RQLGCGSALE AGKEAAFGQG TGPIWLNEVK CKGNETSLWD CPARSWGHSD CGHKEDAAVT CSEIAKSRES LHATGRSHHH HHHHH | |
Greater than 95.0% as determined by: (a) Analysis by HPLC. (b) Analysis by SDS-PAGE. |