missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pig Trypsin Protein
A DNA sequence encoding the Sus scrofa (Pig) Trypsin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9350-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
Trypsin Protein | |
Research Use Only | |
Full Length of Mature Protein | |
Pig | |
His | |
Tris-based buffer, 50% glycerol |
100 μg | |
Not determined. | |
Recombinant Protein | |
Yeast | |
IVGGYTCAAN SIPYQVSLNS GSHFCGGSLI NSQWVVSAAH CYKSRIQVRL GEHNIDVLEG NEQFINAAKI ITHPNFNGNT LDNDIMLIKL SSPATLNSRV ATVSLPRSCA AAGTECLISG WGNTKSSGSS YPSLLQCLKA PVLSDSSCKS SYPGQITGNM ICVGFLEGGK DSCQGDSGGP VVCNGQLQGI VSWGYGCAQK NKPGVYTKVC NYVNWIQQTI AAN | |
Greater than 90% as determined by SDS-PAGE. |