missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pig Transcobalamin-1 Protein
A DNA sequence encoding the Sus scrofa (Pig) Transcobalamin-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9423-iv-100ug
Additional Details : Weight : 0.01000kg
Specifications
Transcobalamin-1 Protein | |
Research Use Only | |
TCN1 | |
Recombinant Protein | |
E. coli | |
CVVSEKDYSH LRLLISAMDN LEQIRGIYGA SILLSQRLAG IQNPSLEEEL SQRIQDDMNR RDMSNLTSGQ LALIILAFGA CKTPDVRFIH DHHLVEKLGE KFKEEIKNME IHNSNPLTNY YQLSFDVLTL CLFRGNYSIS NVTHYFNPEN KNFNLSGHFS VDTGAVAVLA LTCVKRSISN GKIKAAIKDS DTIQKYIESL VHKIQSEKMV VSLETRIAQE KLCRLSLSHQ TITKMNQIAK KLWTRCLTHS QGVFRLPIAA AQILPALLGK TYLDVTKLLL VPKVQVNITD EPVPVVPTLS PENISVIYCV KINEISNCIN ITVFLDVMKA AQEKNSTIYG FTMTETPWGP YITSVQGIWA NNNERTYWEH SEQQQITKPR SMGIMLSKME SI | |
Greater than 80% as determined by SDS-PAGE. |
100 μg | |
Not determined. | |
Full Length of Mature Protein | |
Pig | |
His | |
Tris-based buffer, 50% glycerol |