missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Pig ETFDH Protein

A DNA sequence encoding the Sus scrofa (Pig) ETFDH, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6003-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15939279

  • £2180.00 / 1mg

Estimated Shipment
05-05-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

ETFDH Protein
1 mg
Not determined.
Full Length
Pig
His-SUMO
Tris-based buffer, 50% glycerol
Greater than 85.3% as determined by SDS-PAGE.
Research Use Only
ETFDH
Recombinant Protein
E. coli
SSTCKVPRIT THYTIYPRDQ DKRWEGVNME RFAEEADVVI VGAGPAGLSA ATRLKQLAAQ HEKDLRVCLV EKAAHIGAHT LSGACLDPRA FEELFPDWKE KGAPLNTPVT EDRFGILTEK YRIPVPILPG LPMNNHGNYV VRLGHLVSWM GEQAEALGVE VYPGYAAAEI LFHEDGSVKG IATNDVGIQK DGAPKTTFER GLELHAKVTI FAEGCHGHLA KQLYKKFDLR ANCEPQTYGI GLKELWVIDE KKWKPGRVDH TVGWPLDRHT YGGSFLYHLN EGEPLLALGF VVGLDYQNPY LSPFREFQRW KHHPSIKPTL EGGKRIAYGA RALNEGGFQS IPKLTFPGGL LIGCSPGFMN VPKIKGTHTA MKSGTLAAES IFNQLTSENL QSKTIGLHVT EYEDNLKNSW VWKELYSVRN IRPSCHGILG VYGGMIYTGI FYWIFRGMEP WTLKHKGSDS DQLKPAKDCT PIEYPKPDGQ ISFDLLSSVA LSGTNHEHDQ PAHLTLKDDS VPVNRNLSIY DGPEQRFCPA GVYEFVPLEQ GDGFRLQINA QNCVHCKTCD IKDPSQNINW VVPEGGGGPA YNGM
Videos
SDS
Documents