missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Parvovirus Parvovirus VP2 Protein
Recombinant Protein
£272.00 - £2765.00
Specifications
Name | Parvovirus Parvovirus VP2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Untested |
Product Type | Recombinant Protein |
Cross Reactivity | Parvovirus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15954018
|
enQuireBio™
QP10489-500UG |
500 μg |
£945.00
500µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15944018
|
enQuireBio™
QP10489-2MG |
2 mg |
£2765.00
2mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15934018
|
enQuireBio™
QP10489-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Parvovirus Parvovirus VP2 Protein | |
Untested | |
Parvovirus | |
Native | |
>90% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Human | |
Tris NaCl pH7.9. |