missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Ovine Mutant Prolactin Antagonist Protein

A cDNA sequence encoding the Mutant Prolactin Antagonist was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP14011-EC-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15945409

  • £98.39 / 10µg
Estimated Shipment: 10-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Mutant Prolactin Antagonist Protein
10 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
Ovine Prolactin was lyophilized from a concentrated (1 mg/ml) solution with 0.02%-0.03% NaHCO3.
Greater than 99.0% as determined by Gel Filtration & SDS-PAGE.
Research Use Only
Ovine Prolactin Antagonist mutant form is devoid of agonistic activity and capable of inhibiting biological activity of oPRL or other lactogenic hormones as evidenced by proliferation assay of Nb2 or other cells. The truncated form is more potent inhibitor.
Ovine
His
Videos
SDS
Documents
Certificates
Special Offers

Special Offers