missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Ovine IFN tau Protein
A cDNA sequence encoding the IFN tau was constructed and used to recombinantly synthesize the protein.
£99.15 - £4500.00
Specifications
Gene ID (Entrez) | 100144750 |
---|---|
Name | IFN tau Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | IFNT1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15974618
|
enQuireBio™
QP10676-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15954618
|
enQuireBio™
QP10676-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15964618
|
enQuireBio™
QP10676-1MG |
1 mg |
£4500.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
100144750 | |
Research Use Only | |
IFNT1 | |
Recombinant Protein | |
E. coli | |
MAFVLSLLMA LVLVSYGPGG SLGCYLSRKL MLDARENLKL LDRMNRLSPH SCLQDRKDFG LPQEMVEGDQ LQKDQAFPVL YEMLQQSFNL FYTEHSSAAW DTTLLEQLCT GLQQQLDHLD TCRGQVMGEE DSELGNMDPI VTVKKYFQGI YDYLQEKGYS DCAWEIVRVE MMRALTVSTT LQKRLTKMGG DLNSP | |
Greater than 95.0% as determined by both:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
IFN tau Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The specific activity as determined in a viral resistance assay using bovine kidney MDBK cells was found to be 10,000,000IU/mg. | |
Ovine | |
Untagged | |
Lyophilized from (1 mg/ml) solution containing PBS pH 7.4. |