missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Ovine IFN tau Protein
A cDNA sequence encoding the IFN tau was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10676-2ug
Additional Details : Weight : 0.01000kg
Specifications
100144750 | |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The specific activity as determined in a viral resistance assay using bovine kidney MDBK cells was found to be 10,000,000IU/mg. | |
Ovine | |
Untagged | |
Lyophilized from (1 mg/ml) solution containing PBS pH 7.4. |
IFN tau Protein | |
Research Use Only | |
IFNT1 | |
Recombinant Protein | |
E. coli | |
MAFVLSLLMA LVLVSYGPGG SLGCYLSRKL MLDARENLKL LDRMNRLSPH SCLQDRKDFG LPQEMVEGDQ LQKDQAFPVL YEMLQQSFNL FYTEHSSAAW DTTLLEQLCT GLQQQLDHLD TCRGQVMGEE DSELGNMDPI VTVKKYFQGI YDYLQEKGYS DCAWEIVRVE MMRALTVSTT LQKRLTKMGG DLNSP | |
Greater than 95.0% as determined by both:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |