missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other TXN2 Yeast Protein
A cDNA sequence encoding the TXN2 Yeast was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10903-1mg
Additional Details : Weight : 0.01000kg
Specifications
TXN2 Yeast Protein | |
Research Use Only | |
TXN2 activity is assayed by measuring the change in absorbance at 650 nm at 25C The specific activity was found to be 3 units/mg. | |
Other | |
Untagged | |
Each mg of TRX2 protein contains 20mM phosphate buffer pH 7.4. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MVTQLKSASE YDSALASGDK LVVVDFFATW CGPCKMIAPM IEKFAEQYSD AAFYKLDVDE VSDVAQKAEV SSMPTLIFYK GGKEVTRVVG ANPAAIKQAI ASNV | |
Greater than 95.0% as determined by SDS-PAGE. |