missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Treponema p17 22kDa Protein
A cDNA sequence encoding the Treponema p17 22kDa was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13810-1mg
Additional Details : Weight : 0.01000kg
Specifications
Treponema p17 22kDa Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Treponema p17 22kDa protein solution contains PBS 50mM arginine. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Treponema p17 22kDa is >95% pure as determined by 10% PAGE (coomassie staining). |