missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Treponema p17 22kDa Protein

A cDNA sequence encoding the Treponema p17 22kDa was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13810-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15934779

  • £173.00 / 100µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Treponema p17 22kDa Protein
Research Use Only
Recombinant Protein
E. coli
Treponema p17 22kDa protein solution contains PBS 50mM arginine.
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
Treponema p17 22kDa is >95% pure as determined by 10% PAGE (coomassie staining).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers