missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other TP 5 Protein
A cDNA sequence encoding the TP 5 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13789-100mg
Additional Details : Weight : 0.01000kg
Specifications
TP 5 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The protein (1 mg/ml) was lyophilized with no additives. |
100 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Greater than 99.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |