missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other T. pallidum p41 Protein
A cDNA sequence encoding the T. pallidum p41 was constructed and used to recombinantly synthesize the protein.
£173.00 - £1100.00
Specifications
Name | T. pallidum p41 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15944339
|
enQuireBio™
QP13673-100UG |
100 μg |
£173.00
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15964339
|
enQuireBio™
QP13673-500UG |
500 μg |
£660.00
500µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15954339
|
enQuireBio™
QP13673-1MG |
1 mg |
£1100.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
T. pallidum p41 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Treponema pallidum protein is >90% pure as determined by 10% PAGE (coomassie staining). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
10mM Tris-HCl pH 8.0, 1mM EDTA, 1mMDTT and 8M urea. |