missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Streptavidin Protein
A cDNA sequence encoding the Streptavidin was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13625-HIS-5ug
Additional Details : Weight : 0.01000kg
Specifications
Streptavidin Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRKIVVAAIA VSLTTVSITA SASADPSKDS KAQVSAAEAG ITGTWYNQLG STFIVTAGAD GALTGTYESA VGNAESRYVL TGRYDSAPAT DGSGTALGWT VAWKNNYRNA HSATTWSGQY VGGAEARINT QWLLTSGTTE ANAWKSTLVG HDTFTKVKPS AASIDAAKKA GVNNGNPLDA VQQ |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Lyophilized (1 mg/ml) in 50mM NaCl, pH 9.0. |