missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Streptavidin Protein
A cDNA sequence encoding the Streptavidin was constructed and used to recombinantly synthesize the protein.
£99.15 - £3855.00
Specifications
Name | Streptavidin Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15954189
|
enQuireBio™
QP13625-HIS-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15944189
|
enQuireBio™
QP13625-HIS-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15934189
|
enQuireBio™
QP13625-HIS-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15914189
|
enQuireBio™
QP13625-2MG |
2 mg |
£99.15
2mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15924189
|
enQuireBio™
QP13625-5MG |
5 mg |
£99.15
5mg |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15984179
|
enQuireBio™
QP13625-10MG |
10 mg |
£155.00
10mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15904189
|
enQuireBio™
QP13625-20MG |
20 mg |
£155.00
20mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15974179
|
enQuireBio™
QP13625-100MG |
100 mg |
£1210.00
100mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15994179
|
enQuireBio™
QP13625-1GR |
1 g |
£3855.00
1g |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Streptavidin Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Lyophilized (1 mg/ml) in 50mM NaCl, pH 9.0. |
Research Use Only | |
Recombinant Protein | |
MRKIVVAAIA VSLTTVSITA SASADPSKDS KAQVSAAEAG ITGTWYNQLG STFIVTAGAD GALTGTYESA VGNAESRYVL TGRYDSAPAT DGSGTALGWT VAWKNNYRNA HSATTWSGQY VGGAEARINT QWLLTSGTTE ANAWKSTLVG HDTFTKVKPS AASIDAAKKA GVNNGNPLDA VQQ |