missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other SST Protein
A cDNA sequence encoding the SST was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13598-10mg
Additional Details : Weight : 0.01000kg
Specifications
6750 | |
10 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
Ala-Gly-Cys-Lys-Asn-Phe- Phe-Trp-Lys-Thr-Phe-Thr-Ser-Cys-OH | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
SST Protein | |
Research Use Only | |
SST | |
Other | |
Untagged | |
The protein was lyophilized with no additives. |