missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other SIV p55 Protein
A cDNA sequence encoding the SIV p55 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13512-10ug
Additional Details : Weight : 0.01000kg
Specifications
SIV p55 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
SIV p55 Protein solution containing 10mM Tris, pH 8.0, 140mM NaCl and 400mM L-Arginine. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Greater than 90.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |