missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Protein-G His Protein
A cDNA sequence encoding the Protein-G His was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13167-10mg
Additional Details : Weight : 0.01000kg
Specifications
Protein-G His Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Lyophilized with no additives. |
10 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Greater than 95.0% as determined by SDS-PAGE and HPLC analyses. |