missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other protein 1
A cDNA sequence encoding the Protein A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10853-EC-100mg
Additional Details : Weight : 0.01000kg
Specifications
protein 1 | |
Research Use Only | |
spa | |
Recombinant Protein | |
E. coli | |
MKKKNIYSIR KLGVGIASVT LGTLLISGGV TPAANAAQHD EAQQNAFYQV LNMPNLNADQ RNGFIQSLKD DPSQSANVLG EAQKLNDSQA PKADAQQNKF NKDQQSAFYE ILNMPNLNEE QRNGFIQSLK DDPSQSTNVL GEAKKLNESQ APKADNNFNK EQQNAFYEIL NMPNLNEEQR NGFIQSLKDD PSQSANLLAE AKKLNESQAP KADNKFNKEQ QNAFYEILHL PNLNEEQRNG FIQSLKDDPS QSANLLAEAK KLNDAQAPKA DNKFNKEQQN AFYEILHLPN LTEEQRNGFI QSLKDDPSVS KEILAEAKKL NDAQAPKEED NNKPGKEDGN KPGKEDGNKP GKEDNKKPGK EDGNKPGKED NKKPGKEDGN KPGKEDGNKP GKEDGNKPGK EDGNKPGKED GNGVHVVKPG DTVNDIAKAN GTTADKIAAD NKLADKNMIK PGQELVVDKK QPANHADANK AQALPETGEE NPFIGTTVFG GLSLALGAAL LAGRRREL | |
Greater than 98.0% as determined by SDS-PAGE & RP-HPLC. |
100 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
7 mg human IgG/mg protein-A. | |
Other | |
Untagged | |
The protein was lyophilized from a concentrated (1 mg/ml) solution containing no additives. |