missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other PMSG Protein

A cDNA sequence encoding the PMSG was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13081-1000IU

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15952449

  • £98.39 / 1000 units
Estimated Shipment: 20-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

PMSG Protein
Research Use Only
Recombinant Protein
Native Protein
The PMSG was lyophilized with no additives.
1000 IU
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Videos
SDS
Documents
Certificates
Special Offers

Special Offers