missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Phospholipase A2 Protein

A cDNA sequence encoding the Phospholipase A2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13045-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15942339

  • £1100.00 / 1mg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

406141
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MQVVLGSLFL LLLSTSHGWQ IRDRIGDNEL EERIIYPGTL WCGHGNKSSG PNELGRFKHT DACCRTHDMC PDVMSAGESK HGLTNTASHT RLSCDCDDKF YDCLKNSADT ISSYFVGKMY FNLIDTKCYK LEHPVTGCGE RTEGRCLHYT VDKSKPKVYQ WFDLRKY
Protein is >90% pure as determined by 10% SDS-PAGE.
Phospholipase A2 Protein
Research Use Only
Pla2
Other
Untagged
20mM Tris-HCl, pH 7.2, 1.5M urea and 50% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers