missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other PhI p 5 Protein

A cDNA sequence encoding the PhI p 5 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13042-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15942329

  • £874.00 / 100µg

Estimated Shipment
30-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

PhI p 5 Protein
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
PhI p 5 is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol.
Greater than 80.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
Insect
SVKRSNGSAE VHRGAVPRRG PRGGPGRSYA ADAGYAPATP AAAGAEAGKA TTEEQKLIED INVGFKAAVA AAASVPAADK FKTFEAAFTS SSKAATAKAP GLVPKLDAAY SVAYKAAVGA TPEAKFDSFV ASLTEALRVI AGALEVHAVK PVTEEPGMAK IPAGELQIID KIDAAFKVAA TAAATAPADD KFTVFEAAFN KAIKESTGGA YDTYKCIPSL EAAVKQAYAA TVAAAPQVKY AVFEAALTKA ITAMSEVQKV SQPATGAATV AAGAATTAAG AASGAATVAA GGYKV
Videos
SDS
Documents