missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other PfCs Mosaic Protein
A cDNA sequence encoding the PfCs Mosaic was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13014-100ug
Additional Details : Weight : 0.01000kg
Specifications
PfCs Mosaic Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
150mM Imidazole, pH 8.0, 150mM NaCl, 25mM Sodium Phosphate and 50% glycerol. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC. |