missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other M. Pneumoniae P1-C Protein
A cDNA sequence encoding the M. Pneumoniae P1-C was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12614-100ug
Additional Details : Weight : 0.01000kg
Specifications
M. Pneumoniae P1-C Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Mycoplasma Pneumoniae P1-C is formulated in 1x PBS pH 7.4. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Greater than 95% as determined by 12% PAGE (Coomassie staining). |