missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other M. Pneumoniae P1-C Protein
A cDNA sequence encoding the M. Pneumoniae P1-C was constructed and used to recombinantly synthesize the protein.
£173.00 - £1320.00
Specifications
Name | M. Pneumoniae P1-C Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15940999
|
enQuireBio™
QP12614-100UG |
100 μg |
£173.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15960999
|
enQuireBio™
QP12614-500UG |
500 μg |
£660.00
500µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15950999
|
enQuireBio™
QP12614-1MG |
1 mg |
£1320.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
M. Pneumoniae P1-C Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Greater than 95% as determined by 12% PAGE (Coomassie staining). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Mycoplasma Pneumoniae P1-C is formulated in 1x PBS pH 7.4. |