missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Lassa Capsid Protein
A cDNA sequence encoding the Lassa Capsid was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12538-500ug
Additional Details : Weight : 0.01000kg
Specifications
Lassa Capsid Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Lassa Capsid protein solution contains Phosphate buffered saline and 25mM arginine. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Lassa Capsid is >95% pure as determined by 10% PAGE (coomassie staining). |