missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other JEV Protein
A cDNA sequence encoding the JEV was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12471-1mg
Additional Details : Weight : 0.01000kg
Specifications
JEV Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
20mM Tris-MES pH 6.5, 8M urea, 200mM NaCl and 0.05% Tween-20. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Encephalitis protein is >90% pure as determined by SDS-PAGE (coomassie staining). |