missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Intein Bacillus Circulans Protein

A cDNA sequence encoding the Intein Bacillus Circulans was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12451-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15970459

  • £99.15 / 5µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Intein Bacillus Circulans Protein
Research Use Only
Recombinant Protein
E. coli
MKIEEGKLVI GSLEGCFAKG TNVLMADGSI ECIENIEVGN KVMGKDGRPR EVIKLPRGRE TMYSVVQKSQ HRAHKSDSSR EVPELLKFTC NATHELVVRT PRSVRRLSRT IKGVEYFEVI TFEMGQKKAP DGRIVELVKE VSKSYPISEG PERANELVES YRKASNKAYF EWTIEARDLS LLGSHVRKAT YQTYAPILYE NDHFFDYMQK SKFHLTIEGP KVLAYLLGLW IGDGLSDRAT FSVDSRDTSL MERVTEYAEK LNLCAEYKDR KEPQVAKTVN LYSKVVRGAS TNPGVSAWQV NTAYTAGQLV TYNGKTYKCL QPHTSLAGWE PSNVPALWQL QGGHGGIRNN LNTENPLWDA IVGLGFLKDG VKNIPSFLST DNIGTRETFL AGLIDSDGYV TDEHGIKATI KTIHTSVRDG LVSLARSLGL VVSVNAEPAK VDMNVTKHKI SYAIYMSGGD VLLNVLSKCA GSKKFRPAPA AAFARECRGF YFELQELKED DYYGITLSDD SDHQFLLGSQ VVVQNLEHHH HHH
Greater than 95% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
The Intein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 0.1M NaCl and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers