missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other HPSE WB Protein
Recombinant Protein
Brand: enQuireBio™ QP12305-200ng
Additional Details : Weight : 0.01000kg
Specifications
10855 | |
200 ng | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MLLRSKPALP PPLMLLLLGP LGPLSPGALP RPAQAQDVVD LDFFTQEPLH LVSPSFLSVT IDANLATDPR FLILLGSPKL RTLARGLSPA YLRFGGTKTD FLIFDPKKES TFEERSYWQS QVNQDICKYG SIPPDVEEKL RLEWPYQEQL LLREHYQKKF KNSTYSRSSV DVLYTFANCS GLDLIFGLNA LLRTADLQWN SSNAQLLLDY CSSKGYNISW ELGNEPNSFL KKADIFINGS QLGEDFIQLH KLLRKSTFKN AKLYGPDVGQ PRRKTAKMLK SFLKAGGEVI DSVTWHHYYL NGRTATKEDF LNPDVLDIFI SSVQKVFQVV ESTRPGKKVW LGETSSAYGG GAPLLSDTFA AGFMWLDKLG LSARMGIEVV MRQVFFGAGN YHLVDENFDP LPDYWLSLLF KKLVGTKVLM ASVQGSKRRK LRVYLHCTNT DNPRYKEGDL TLYAINLHNV TKYLRLPYPF SNKQVDKYLL RPLGPHGLLS KSVQLNGLTL KMVDDQTLPP LMEKPLRPGS SLGLPAFSYS FFVIRNAKVA ACI |
HPSE WB Protein | |
Research Use Only | |
HPSE | |
Other | |
Untagged | |
Concentration: 1ug /ml Content: 100 ngBuffer: LDS-PAGE buffer[140 mM Tris buffer pH 8.5, 10% Glycerol, 2% LDS, 0.015% EDTA, 1.88% (v/v) of 1% Serva Blue G250 and 0.625% (v/v) of 1% Phenol red]. |