missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other GFP Protein
A cDNA sequence encoding the GFP was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11955-2ug
Additional Details : Weight : 0.01000kg
Specifications
GFP Protein | |
Research Use Only | |
Recombinant Protein | |
Non-species Specific | |
The protein was lyophilized from a 1 mg/ml solution containing 10mM sodium phosphate buffer pH 7.5, 6M urea, 2mM DTT, 1mM EDTA and 10mM methylammonium chloride. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |