missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other GAGA-POZ Protein
A cDNA sequence encoding the GAGA-POZ was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11922-1mg
Additional Details : Weight : 0.01000kg
Specifications
GAGA-POZ Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MSLPMNSLYS LTWGDYGTSL VSAIQLLRCH GDLVDCTLAA GGRSFPAHKI VLCAASPFLLDLLKNTPCKH PVVMLAGVNA NDLEALLEFV YRGEVSVDHA QLPSLLQAAQ CLNIQGLAPQTVTKDDYTTH | |
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
The protein containing 10mM HEPES (pH 7.4) and 25mM NaCl. |