missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Exenatide Protein

A cDNA sequence encoding the Exenatide was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11816-50mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15978468

  • £2535.00 / 50mg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Exenatide Protein
Research Use Only
Recombinant Protein
E. coli
H-His-Gly-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Leu-Ser-Lys-Gln-Met-Glu-Glu-Glu-Ala-Val-Arg-Leu-Phe-Ile-Glu-Trp-Leu-Lys-Asn-Gly-Gly-Pro-Ser-Ser-Gly-Ala-Pro-Pro-Pro-Ser-NH2
Greater than 99.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
50 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
The Exenatide peptide was lyophilized from a concentrated solution with no additives.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers