missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Elcatonin Protein
Recombinant Protein
£99.15 - £2975.00
Specifications
Name | Elcatonin Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | Biological Activity (based on net peptide) was found to be 6695.2 IU/mg. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15954328
|
enQuireBio™
QP10585-200UG |
200 μg |
£99.15
200µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15944328
|
enQuireBio™
QP10585-1MG |
1 mg |
£155.00
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15934328
|
enQuireBio™
QP10585-100MG |
100 mg |
£2975.00
100mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Elcatonin Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Untagged | |
The protein was lyophilized with no additives. |
Research Use Only | |
Biological Activity (based on net peptide) was found to be 6695.2 IU/mg. | |
Other | |
Ser-Asn-Leu-Ser-Thr-Asu-Val-Leu-Gly-Lys-Leu-Ser-Gln-Glu-Leu-His-Lys-Leu-Gln-Thr-Tyr-Pro-Arg-Thr-Asn-Val-Gly-Ala-Gly-Thr-Pro-NH2 | |
Greater than 93.3% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |