Learn More
enQuireBio™ Recombinant other Elcatonin Protein
Recombinant Protein
Brand:  enQuireBio™ QP10585-200ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15954328
  • £99.15 / 200µg
Estimated Shipment: 10-05-2024
to see stock.
Specifications
Elcatonin Protein
Research Use Only
Biological Activity (based on net peptide) was found to be 6695.2 IU/mg.
Other
Ser-Asn-Leu-Ser-Thr-Asu-Val-Leu-Gly-Lys-Leu-Ser-Gln-Glu-Leu-His-Lys-Leu-Gln-Thr-Tyr-Pro-Arg-Thr-Asn-Val-Gly-Ala-Gly-Thr-Pro-NH2
Greater than 93.3% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
200 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
Untagged
The protein was lyophilized with no additives.