missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other DsbE Protein
A cDNA sequence encoding the DsbE was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11705-1mg
Additional Details : Weight : 0.01000kg
Specifications
DsbE Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRNAEGDDPT NLESALIGKP VPKFRLESLD NPGQFYQADV LTQGKPVLLN VWATWCPTCR AEHQYLNQLS AQGIRVVGMN YKDDRQKAIS WLKELGNPYA LSLFDGDGML GLDLGVYGAP ETFLIDGNGI IRYRHAGDLN PRVWEEEIKP LWEKYSKEAA Q | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
The DsbE protein solution contains 20mM Tris-HCl, pH 7.5, 2mM EDTA and 10% Glycerol. |