missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other DsbE Protein
A cDNA sequence encoding the DsbE was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | DsbE Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15958098
|
enQuireBio™
QP11705-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15948098
|
enQuireBio™
QP11705-25UG |
25 μg |
£155.00
25µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15938098
|
enQuireBio™
QP11705-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
DsbE Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
The DsbE protein solution contains 20mM Tris-HCl, pH 7.5, 2mM EDTA and 10% Glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRNAEGDDPT NLESALIGKP VPKFRLESLD NPGQFYQADV LTQGKPVLLN VWATWCPTCR AEHQYLNQLS AQGIRVVGMN YKDDRQKAIS WLKELGNPYA LSLFDGDGML GLDLGVYGAP ETFLIDGNGI IRYRHAGDLN PRVWEEEIKP LWEKYSKEAA Q | |
Greater than 95.0% as determined by SDS-PAGE. |