missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other DnaK Protein
A cDNA sequence encoding the DnaK was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11682-50ug
Additional Details : Weight : 0.01000kg
Specifications
DnaK Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGKIIGIDLG TTNSCVAIMD GTTPRVLENA EGDRTTPSII AYTQDGETLV GQPAKRQAVTNPQNTLFAIK RLIGRRFQDE EVQRDVSIMP FKIIAADNGD AWVEVKGQKM APPQISAEVLKKMKKTAEDY LGEPVTEAVI TVPAYFNDAQ RQATKDAGRI AGLEVKRIINEPTAAALAYGLDKGTGNRTI AVYDLGGGTFDISIIEIDEV DGEKTFEVLA TNGDTHLGGE DFDSRLINYLVEEFKKDQGI DLRNDPLAMQ RLKEAAEKAK IELSSAQQTD VNLPYITADA TGPKHMNIKVTRAKLESLVE DLVNRSIEPL KVALQDAGLS VSDIDDVILV GGQTRMPMVQ KKVAEFFGKEPRKDVNPDEA VAIGAAVQGG VLTGDVKDVL LLDVTPLSLG IETMGGVMTT LIAKNTTIPTKHSQVFSTAE DNQSAVTIHV LQGERKRAAD NKSLGQFNLD GINPAPRGMPQIEVTFDIDADGILHVSAKD KNSGKEQKITIKASSGLNED EIQKMVRDAE ANAEADRKFE ELVQTRNQGDHLLHSTRKQV EEAGDKLPAD DKTAIESALT ALETALKGED KAAIEAKMQE LAQVSQKLMEIAQQQHAQQQ TAGADASANN AKDDDVVDAE FEEVKDKK | |
Greater than 95.0% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
The DnaK protein contains 25mM Tris-HCl buffer (pH 7.5), 100mM NaCl, 5mM DTT and 10% Glycerol. |