missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Cys-Protein-L Protein

A cDNA sequence encoding the Cys-Protein-L was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11588-10mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15967728

  • £151.37 / 10mg
Estimated Shipment: 14-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Cys-Protein-L Protein
10 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Protein-L was lyophilized without any additives.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
CKEETPETPE TDSEEEVTIK ANLIFANGST QTAEFKGTFE KATSEAYAYA DTLKKDNGEY TVDVADKGYT LNIKFAGKEK TPEEPKEEVT IKANLIYADG KTQTAEFKGT FEEATAEAYR YADALKKDNG EYTVDVADKG YTLNIKFAGK EKTPEEPKEE VTIKANLIYA DGKTQTAEFK GTFEEATAEA YRYADLLAKE NGKYTVDVAD KGYTLNIKFA GKEKTPEEPK EEVTIKANLI YADGKTQTAE FKGTFAEATA EAYRYADLLA KENGKYTADL EDGGYTINIR FAGKKVDEKP EEKEQVTIKE NIYFEDGTVQ TATFKGTFAE ATAEAYRYAD LLSKEHGKYT ADLEDGGYTI NIRFAG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers