missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other CMV Mosaic Protein
A cDNA sequence encoding the CMV Mosaic was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11461-100ug
Additional Details : Weight : 0.01000kg
Specifications
CMV Mosaic Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
CMV Mosaic protein solution (0.42 mg/ml) contains Phosphate buffered saline, 2M urea and 100mM arginine. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
CMV Mosaic is >95% pure as determined by 12% PAGE (coomassie staining). |