missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other CK2a Zea Mays Protein

A cDNA sequence encoding the CK2a Zea Mays was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11433-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15947248

  • £1654.24 / 20µg
Estimated Shipment: 14-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CK2a Zea Mays Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
CK2a is supplied in 50% glycerol.
Greater than 99% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MSKARVYADV NVLRPKEYWD YEALTVQWGE QDDYEVVRKV GRGKYSEVFE GINVNNNEKC IIKILKPVKK KKIKREIKIL QNLCGGPNIV KLLDIVRDQH SKTPSLIFEY VNNTDFKVLY PTLTDYDIRY YIYELLKALD YCHSQGIMHR DVKPHNVMID HELRKLRLID WGLAEFYHPG KEYNVRVASR YFKGPELLVD LQDYDYSLDM WSLGCMFAGM IFRKEPFFYG HDNHDQLVKI AKVLGTDGLN VYLNKYRIEL DPQLEALVGR HSRKPWLKFM NADNQHLVSP EAIDFLDKLL RYDHQERLTA LEAMTHPYFQ QVRAAENSRT RA
Videos
SDS
Documents
Certificates
Special Offers

Special Offers