missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other CK2a Zea Mays Protein
A cDNA sequence encoding the CK2a Zea Mays was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11433-10ug
Additional Details : Weight : 0.01000kg
Specifications
CK2a Zea Mays Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MSKARVYADV NVLRPKEYWD YEALTVQWGE QDDYEVVRKV GRGKYSEVFE GINVNNNEKC IIKILKPVKK KKIKREIKIL QNLCGGPNIV KLLDIVRDQH SKTPSLIFEY VNNTDFKVLY PTLTDYDIRY YIYELLKALD YCHSQGIMHR DVKPHNVMID HELRKLRLID WGLAEFYHPG KEYNVRVASR YFKGPELLVD LQDYDYSLDM WSLGCMFAGM IFRKEPFFYG HDNHDQLVKI AKVLGTDGLN VYLNKYRIEL DPQLEALVGR HSRKPWLKFM NADNQHLVSP EAIDFLDKLL RYDHQERLTA LEAMTHPYFQ QVRAAENSRT RA | |
Greater than 99% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
CK2a is supplied in 50% glycerol. |