missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other C. Albicans Bgl2 Protein

A cDNA sequence encoding the C. Albicans Bgl2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11215-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15956508

  • £990.00 / 100µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

C. Albicans Bgl2 Protein
Research Use Only
Recombinant Protein
Baculovirus
C. Albicans Bgl2 is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol.
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
Greater than 80.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers