missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other C. Albicans Bgl2 Protein
A cDNA sequence encoding the C. Albicans Bgl2 was constructed and used to recombinantly synthesize the protein.
£696.00 - £4640.00
Specifications
Name | C. Albicans Bgl2 Protein |
---|---|
Product Type | Recombinant Protein |
Protein Tag | His |
Host Species | Insect |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15976508
|
enQuireBio™
QP11215-50UG |
50 μg |
£696.00
50µg |
Estimated Shipment: 14-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
15956508
|
enQuireBio™
QP11215-100UG |
100 μg |
£1045.00
100µg |
Estimated Shipment: 14-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
15966508
|
enQuireBio™
QP11215-1MG |
1 mg |
£4640.00
1mg |
Estimated Shipment: 14-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
Specifications
C. Albicans Bgl2 Protein | |
His | |
Other |
Recombinant Protein | |
Insect | |
Greater than 80.0% as determined by SDS-PAGE. |