missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other BSA Protein

A cDNA sequence encoding the BSA was constructed and used to recombinantly synthesize the protein.

£196.78 - £481.13

Specifications

Name BSA Protein
Product Type Recombinant Protein
Protein Tag Untagged
Host Species Native Protein
Cross Reactivity Other
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15936478
View Documents Promotion Details
enQuireBio™
QP11206-10GR
10 g £196.78
10g
Qty Price Break Available View alternative products
Save Up to 
»
Estimated Shipment: 02-02-2022
Please to purchase this item. Need a web account? Register with us today!
15946478
View Documents Promotion Details
enQuireBio™
QP11206-50GR
50 g
£320.04
50g
View alternative products
Save Up to 
»
Estimated Shipment: 18-02-2022
Please to purchase this item. Need a web account? Register with us today!
15926478
View Documents Promotion Details
enQuireBio™
QP11206-100GR
100 g
£481.13
100g
View alternative products
Save Up to 
»
Estimated Shipment: 18-02-2022
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

BSA Protein
Untagged
Other
Greater than 97.0%.
ALB
Recombinant Protein
Native Protein
MKWVTFISLL LLFSSAYSRG VFRRDTHKSE IAHRFKDLGE EHFKGLVLIA FSQYLQQCPF DEHVKLVNEL TEFAKTCVAD ESHAGCEKSL HTLFGDELCK VASLRETYGD MADCCEKQEP ERNECFLSHK DDSPDLPKLK PDPNTLCDEF KADEKKFWGK YLYEIARRHP YFYAPELLYY ANKYNGVFQE CCQAEDKGAC LLPKIETMRE KVLASSARQR LRCASIQKFG ERALKAWSVA RLSQKFPKAE FVEVTKLVTD LTKVHKECCH GDLLECADDR ADLAKYICDN QDTISSKLKE CCDKPLLEKS HCIAEVEKDA IPENLPPLTA DFAEDKDVCK NYQEAKDAFL GSFLYEYSRR HPEYAVSVLL RLAKEYEATL EECCAKDDPH ACYSTVFDKL KHLVDEPQNL IKQNCDQFEK LGEYGFQNAL IVRYTRKVPQ VSTPTLVEVS RSLGKVGTRC CTKPESERMP CTEDYLSLIL NRLCVLHEKT PVSEKVTKCC TESLVNRRPC FSALTPDETY VPKAFDEKLF TFHADICTLP DTEKQIKKQT ALVELLKHKP KATEEQLKTV MENFVAFVDK CCAADDKEAC FAVEGPKLVV STQTALA
280717
SDS
Product Certifications