missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Borrelia Garinii p58 Protein
A cDNA sequence encoding the Borrelia Garinii p58 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11185-1mg
Additional Details : Weight : 0.01000kg
Specifications
Borrelia Garinii p58 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Borrelia Garinii p58 is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Greater than 80.0% as determined by SDS-PAGE. |